Merck
Cell-free plasma DNA as a novel marker of aseptic inflammation severity related to exercise overtraining.
Clinical chemistry (2006-07-15)
Ioannis G Fatouros, Aspasia Destouni, Konstantinos Margonis, Athanasios Z Jamurtas, Christina Vrettou, Dimitrios Kouretas, George Mastorakos, Asimina Mitrakou, Kiriakos Taxildaris, Emmanouel Kanavakis, Ioannis Papassotiriou
ABSTRACT
Circulating free plasma DNA is implicated in conditions associated with tissue injury, including exercise-induced inflammation, and thus is a potential marker for athletic overtraining. We measured free plasma DNA along with C-reactive protein (CRP), creatine kinase (CK), and uric acid (UA) in 17 recreationally trained men participating in a 12-week resistance training regimen (8 resistance multi-joint exercises selected to stress the entire musculature: bench press, squat, leg press, snatch, hang clean, dead lifts, barbell arm curls, and rowing), consisting of 4 training periods (t1, t2, t3, and t4). Plasma DNA concentrations increased markedly after t1, t2, and t3 and returned to baseline after t4. There were substantial differences between t2 and t1 and between t3 and t2 plasma DNA concentrations. CRP increased by 300% after t2 and by 400% after t3 (there was no difference between t2 and t3 CRP values) compared with baseline (t0). CK increased only after t3. UA increased after t2 and t3, with a greater increase after t3. This study demonstrates that, after chronic excessive resistance exercise, plasma DNA concentrations increase in proportion to training load, suggesting that plasma DNA may be a sensitive marker for overtraining-induced inflammation.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Roche
Human Genomic DNA, from human blood (buffy coat)