Merck
  • Comparative protection against liver inflammation and fibrosis by a selective cyclooxygenase-2 inhibitor and a nonredox-type 5-lipoxygenase inhibitor.
Comparative protection against liver inflammation and fibrosis by a selective cyclooxygenase-2 inhibitor and a nonredox-type 5-lipoxygenase inhibitor.
The Journal of pharmacology and experimental therapeutics (2007-09-04)
Raquel Horrillo, Anna Planagumà, Ana González-Périz, Natàlia Ferré, Esther Titos, Rosa Miquel, Marta López-Parra, Jaime L Masferrer, Vicente Arroyo, Joan Clària
ABSTRACT
In this study, we examined the relative contribution of cyclooxygenase-2 (COX-2) and 5-lipoxygenase (5-LO), two major proinflammatory pathways up-regulated in liver disease, to the progression of hepatic inflammation and fibrosis. Separate administration of 4-[5-(4-chlorophenyl)-3-(trifluoromethyl)-1H-pyrazol-1-yl]benzenesulfonamide (SC-236), a selective COX-2 inhibitor, and CJ-13,610, a 5-LO inhibitor, to carbon tetrachloride-treated mice significantly reduced fibrosis as revealed by the analysis of Sirius Red-stained liver sections without affecting necroinflammation. Conversely, combined administration of SC-236 and 4-[3-[4-(2-methylimidazol-1-yl)-phenylthio]]phenyl-3,4,5,6-tetrahydro-2H-pyran-4-carboxamide (CJ-13,610) reduced both necroinflammation and fibrosis. These findings were confirmed in 5-LO-deficient mice receiving SC-236, which also showed reduced hepatic monocyte chemoattractant protein 1 expression. Interestingly, SC-236 and CJ-13,610 significantly increased the number of nonparenchymal liver cells with apoptotic nuclei (terminal deoxynucleotidyl transferase dUTP nick-end labeling-positive). Additional pharmacological profiling of SC-236 and CJ-13,610 was performed in macrophages, the primary hepatic inflammatory cell type. In these cells, SC-236 inhibited prostaglandin (PG) E2 formation in a concentration-dependent manner, whereas CJ-13,610 blocked leukotriene B4 biosynthesis. Of note, the simultaneous addition of SC-236 and CJ-13,610 resulted in a higher inhibitory profile on PGE2 biosynthesis than the dual COX/5-LO inhibitor licofelone. These drugs differentially regulated interleukin-6 mRNA expression in macrophages. Taken together, these findings indicate that both COX-2 and 5-LO pathways are contributing factors to hepatic inflammation and fibrosis and that these two pathways of the arachidonic acid cascade represent potential targets for therapy.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
CJ-13610, ≥98% (HPLC)