Merck
  • Polybrominated diphenyl ethers (PBDEs) and alternative brominated flame retardants in air and seawater of the European Arctic.
Polybrominated diphenyl ethers (PBDEs) and alternative brominated flame retardants in air and seawater of the European Arctic.
Environmental pollution (Barking, Essex : 1987) (2011-03-23)
Axel Möller, Zhiyong Xie, Renate Sturm, Ralf Ebinghaus
ABSTRACT
The spatial distribution of polybrominated diphenyl ethers (PBDEs) and several alternative non-PBDE, non-regulated brominated flame retardants (BFRs) in air and seawater and the air-seawater exchange was investigated in East Greenland Sea using high-volume air and water samples. Total PBDE concentrations (Ó₁₀PBDEs) ranged from 0.09 to 1.8 pg m⁻³ in the atmosphere and from 0.03 to 0.64 pg L⁻¹ in seawater. Two alternative BFRs, Hexabromobenzene (HBB) and 2,3-dibromopropyl-2,4,6-tribromophenyl ether (DPTE), showed similar concentrations and spatial trends as PBDEs. The air-seawater gas exchange was dominated by deposition with fluxes up to -492 and -1044 pg m⁻² day⁻¹ for BDE-47 and DPTE, respectively. This study shows the first occurrence of HBB, DPTE and other alternative flame retardants (e.g., pentabromotoluene (PBT)) in the Arctic atmosphere and seawater indicating that they have a similar long-range atmospheric transport potential (LRAT) as the banned PBDEs.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Hexabromobenzene, 98%