Merck
Atrophy and Death of Nonpeptidergic and Peptidergic Nociceptive Neurons in SIV Infection.
The American journal of pathology (2020-04-05)
Jake A Robinson, Guy Guenthner, Rebecca Warfield, Jessica R Kublin, Mandy D Smith, Masoud Shekarabi, Andrew D Miller, Tricia H Burdo
ABSTRACT
HIV-associated sensory neuropathy is a common neurologic comorbidity of HIV infection and prevails in the post-antiretroviral therapy (ART) era. HIV infection drives pathologic changes in the dorsal root ganglia (DRG) through inflammation, altered metabolism, and neuronal dysfunction. Herein, we characterized specific neuronal populations in an SIV-infected macaque model with or without ART. DRG neuronal populations were identified by neurofilament H-chain 200, I-B4 isolectin (IB4), or tropomyosin receptor kinase A expression and assessed for cell body diameter, population size, apoptotic markers, and regeneration signaling. IB4+ and tropomyosin receptor kinase A-positive neurons showed a reduced cell body size (atrophy) and decreased population size (cell death) in the DRG of SIV-infected animals compared with uninfected animals. IB4+ nonpeptidergic neurons were less affected in the presence of ART. DRG neurons showed accumulation of cleaved caspase 3 (apoptosis) and nuclear-localized activating transcription factor 3 (regeneration) in SIV infection, which was significantly lower in uninfected animals and SIV-infected animals receiving ART. Nonpeptidergic neurons predominantly colocalized with cleaved caspase 3 staining. Nonpeptidergic and peptidergic neurons colocalized with nuclear-accumulated activating transcription factor 3, showing active regeneration in sensory neurons. These data suggest that nonpeptidergic and peptidergic neurons are susceptible to pathologic changes from SIV infection, and intervention with ART did not fully ameliorate damage to the DRG, specifically to peptidergic neurons.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
Anti-ATF3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anticorpo anti-neurofilamento di 200 kDa, clone N52, clone N52, Chemicon®, from mouse