Merck
  • A Single-Ion Conducting Borate Network Polymer as a Viable Quasi-Solid Electrolyte for Lithium Metal Batteries.
A Single-Ion Conducting Borate Network Polymer as a Viable Quasi-Solid Electrolyte for Lithium Metal Batteries.
Advanced materials (Deerfield Beach, Fla.) (2020-01-28)
Dong-Myeong Shin, Jonathan E Bachman, Mercedes K Taylor, Jovan Kamcev, Jesse G Park, Michael E Ziebel, Ever Velasquez, Nanette N Jarenwattananon, Gurmukh K Sethi, Yi Cui, Jeffrey R Long
ABSTRACT
Lithium-ion batteries have remained a state-of-the-art electrochemical energy storage technology for decades now, but their energy densities are limited by electrode materials and conventional liquid electrolytes can pose significant safety concerns. Lithium metal batteries featuring Li metal anodes, solid polymer electrolytes, and high-voltage cathodes represent promising candidates for next-generation devices exhibiting improved power and safety, but such solid polymer electrolytes generally do not exhibit the required excellent electrochemical properties and thermal stability in tandem. Here, an interpenetrating network polymer with weakly coordinating anion nodes that functions as a high-performing single-ion conducting electrolyte in the presence of minimal plasticizer, with a wide electrochemical stability window, a high room-temperature conductivity of 1.5 × 10-4 S cm-1 , and exceptional selectivity for Li-ion conduction (tLi+ = 0.95) is reported. Importantly, this material is also flame retardant and highly stable in contact with lithium metal. Significantly, a lithium metal battery prototype containing this quasi-solid electrolyte is shown to outperform a conventional battery featuring a polymer electrolyte.
MATERIALI
N° Catalogo
Marchio
Descrizione del prodotto
Sigma-Aldrich
cis-2-Butene-1,4-diol, 97%