Documenti fondamentali
55087-U
ValvTool
About This Item
Codice UNSPSC:
41115854
NACRES:
SB.51
Descrizione
5/16 in. socket × 1/4 in. open wrench
Descrizione generale
1/4
The ValvTool is a uniquely designed wrench that provides easy access for changing sample loops or replacing fittings on Rheodyne valves. The open end can be used for working with in. stainless steel HPLC fittings, as well.
Prodotti consigliati
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)