Documenti fondamentali
502693
Upchurch Precolumn MicroFilter
for use with 1/16 in. O.D. tubing, PEEK frit
About This Item
Codice UNSPSC:
41115700
Materiali
PEEK frit
Confezionamento
pkg of 1 ea
Raccordi
10-32 female Upchurch inlet fitting for 1/16 in. O.D. tubing
10-32 male Upchurch outlet fitting for 1/16 in.
Compatibilità
for use with 1/16 in. O.D. tubing
Descrizione generale
PEEK housing with in-line filter connects capillary tubing (using Microtight tubing sleeves) or 1/16 in. O.D. tubing to female 10-32 fitting. Swept volume of 0.3 μL. Includes 5 × 0.5 μm frits (one installed).
Prodotti consigliati
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 2
1 of 2
Frit for Supelco® Filter stainless steel, pore size 0.5 μm, pkg of 5 ea
Z290874
Frit for Supelco® Filter
PEEK Tubing L × O.D. × I.D. 10 ft × 1/16 in. × 0.005 in., red, max. pressure 8000
Z227307
PEEK Tubing