22067
Swagelok® Connector to Male NPT
stainless steel, 1/8 in. Swagelok, 1/4 in. male NPT, 200-1-4
Sinonimo/i:
Swagelok® 200-1-4,SS CONNECTOR 1/8TX1/4P EA
About This Item
Prodotti consigliati
Materiali
stainless steel
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
200-1-4
Raccordi
1/4 in. male NPT
1/8 in. Swagelok
Descrizione generale
Nuts and ferrules included.
Note legali
Swagelok is a registered trademark of Swagelok Company
Choose from one of the most recent versions:
Certificati d'analisi (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Servizio Clienti