Documenti fondamentali
2049801
Supelco
Inlet liner, split with cup (unpacked) for Varian® 1075/1077 injector
About This Item
Codice UNSPSC:
41121800
Materiali
glass
Descrizione
Inlet Liner, Split with cup, for Varian
Inlet liner
cup yes unpacked
Caratteristiche
deactivation high temperature silanization
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Varian 01-900109-02 (OEM)
Lungh. × D.E. × D.I.
72 mm × 6.3 mm × 4 mm
Lungh. × D.E. liner
72 mm × 6.3 mm
Compatibilità
configured for
for use with Varian 1075/1077 Injectors
for use with Varian
technique used for High & low MW analytes
Note legali
Varian is a registered trademark of Varian Medical Systems, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)