Z762768
Eppendorf® Combitips advanced® pipette tips, Biopur® grade
maximum volume 0.5 mL
Sinonimo/i:
pipetting tips
About This Item
Codice UNSPSC:
41121600
NACRES:
NB.01
Prodotti consigliati
Materiali
purple tip
Sterilità
sterile
Caratteristiche
barrier: no
Confezionamento
pack of 100 ea individually wrapped
Produttore/marchio commerciale
Eppendorf® 0030089634
Volume massimo
0.5 mL
Descrizione generale
The Combitips advanced are optimized for all Repeater models to create the perfect connection!
The new Eppendorf Combitips advanced feature the characteristic Eppendorf 3D design on the upper rim of the Combitip. With 9 volume sizes (0.1 mL to 50 mL) and Eppendorf Quality, Sterile, PCR Clean and Eppendorf Biopur purity grades, you will always find the perfect Combitip for your application.
The "Biopur" tips are individually blister-wrapped and feature an access tab which makes them easier to open, even with gloves.
The new Eppendorf Combitips advanced feature the characteristic Eppendorf 3D design on the upper rim of the Combitip. With 9 volume sizes (0.1 mL to 50 mL) and Eppendorf Quality, Sterile, PCR Clean and Eppendorf Biopur purity grades, you will always find the perfect Combitip for your application.
The "Biopur" tips are individually blister-wrapped and feature an access tab which makes them easier to open, even with gloves.
Caratteristiche e vantaggi
- pyrogen-free
- free of human DNA
- free of bacterial DNA
- RNase-free
- PCR inhibitor-free
- DNase-free
- ATP-free
Note legali
Biopur is a registered trademark of Eppendorf AG
Combitips advanced is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)