Documenti fondamentali
Z757721
Sartorius pipette tips
volume range 0.5-350 μL, standard, refill, sterile
Scegli un formato
1440 EA
CHF 120.00
CHF 120.00
Spedizione prevista il22 maggio 2025
Scegli un formato
Cambia visualizzazione
1440 EA
CHF 120.00
About This Item
Codice UNSPSC:
41121600
NACRES:
NB.01
CHF 120.00
Spedizione prevista il22 maggio 2025
Materiali
colorless
Sterilità
sterile
Caratteristiche
DNase free
RNase free
barrier: no
endotoxin free
Confezionamento
pack of 96 ea (Color coded tip trays)
Produttore/marchio commerciale
Sartorius 790353
Volume massimo
350 μL
Intervallo di volume
0.5-350 μL
Volume
5—350 μL
Descrizione generale
The non-filtered Optifit Tips are designed for best possible fitting and sealing to secure the highest possible accuracy and precision. These pipette tips enable ergonomic and light tip attachment.
Optifit Tips are available in various packaging types for all your needs:
  • Single Tray racks – purity certified tip racks
  • Refill Packs - purity certified and ecological refill packages
  • Refill Towers - space-saving and ecological refill packages
  • Flexibulk boxes - space-saving, ecological and economical bulk boxes
  • Standard bulk – economical bulk boxes
While Optifit Tip trays are colour coded for easy matching with a corresponding, colour coded Sartorius pipette, they do also match with most other pipette due to their universal design.
Strict quality standards, ISO 9001 and ISO 13485, are followed from tip R&D to production and delivery. The purity certified tips are manufactured in ISO 8 classified clean room conditions to avoid any contaminations and are tested for DNase, RNase and endotoxins per each tip lot. Pre-sterilised tips are also available for the purest laboratory work. Sartorius offers a selection of pipette tips, extended and large volume tips, for special needs.
Altre note
Browse all pipette tips
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)