Documenti
Z757551
Sartorius eLINE® single channel electronic pipettor
volume 50-1000 μL, without adapter
Sinonimo/i:
Biohit pipettes, electronic pipettes, pipettor
About This Item
Codice UNSPSC:
41121500
Ingresso CA/CC
100 V / 240 V AC, European 2-pin plug (flat 2-pin)
100 V / 240 V AC, UK plug (3 pin)
100 V / 240 V AC, US 2-pin plug
Caratteristiche
CE compliant
CSA approved: no
electronic
single-channel
Produttore/marchio commerciale
Sartorius 730080
Volume
50-1000 μL
Descrizione generale
Sartorius eLINE electronic pipettors of the Biohitfamily offer the latest technology in liquid handling. The eLINE electronic pipettor has been designed to deliver the highest levels of analytical accuracy with minimal the eLINE pipettors together with the ejection - take the effort out of pipetting in every laboratory.
The eLINE modes including:
  • Forward Pipetting with Optional Cycle
  • Reverse Pipetting
  • Manual Pipetting
  • Post Delivery Mixing
  • Multi-Dispensing
  • Multi-Aspirating
  • Automated Multi-Dispensing
  • Sequential-Dispensing
Note legali
eLINE is a registered trademark of Sartorius Biohit Liquid Handling Oy