Documenti fondamentali
Z742664
Molecular Grinding Resin
with Pestle & Tubes
Scegli un formato
100 EA
CHF 350.00
CHF 350.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
100 EA
CHF 350.00
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.74
CHF 350.00
Check Cart for Availability
Caratteristiche
with Pestle & Tubes
Confezionamento
pkg of 100 ea
Produttore/marchio commerciale
G-BioSciences 786-138PR
Categorie correlate
Descrizione generale
Includes:
Molecular Grinding Resin 5 x 0.5 ml resin
Grinding Pestles (RNase Free) 100 x
Grinding Tubes (RNase Free) 100 x
Molecular Grinding Resin has been specifically developed for effectively grinding biological samples for extraction of DNA, RNA and proteins. The resin is high tensile micro-particles, which effectively disrupt nuclei and other cell organelles. Molecular Grinding Resin does not bind protein or nucleic acids. Simply mix the biological sample with Molecular Grinding Resin and grind or homogenize the sample. Unlike sand or other grinding materials, Molecular Grinding Resin is easy to work with and does not damage genomic DNA or high molecular mass proteins. We routinely use Molecular Grinding Resin for isolation of 100kb genomic DNA and RNA. Molecular Grinding Resin can be used with any homogenization technique, including high speed mechanical grinding such as Polytron and high-energy sonicators. Molecular Grinding Resin is also available with disposable matching pestles and tubes.
Note legali
Molecular Grinding Resin is a trademark of Geno Technology, Inc.
Prodotti correlati
N° Catalogo
Descrizione
Determinazione del prezzo
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 3
1 of 3
Dounce tissue grinder tube working volume 2 mL
T2690
Dounce tissue grinder tube