Each pack contains 1000 labels.
Z740259
Tough-Spots®
size 3/8 in., red
Sinonimo/i:
solvent resistant labels, temperature resistant labels
Scegli un formato
CHF 34.00
Scegli un formato
About This Item
CHF 34.00
Prodotti consigliati
Materiali
PVC
red
Confezionamento
pkg of 1000 ea (per roll)
Produttore/marchio commerciale
Diversified Biotech T-SPOTS-R
Misura
3/8 in.
Categorie correlate
Descrizione generale
Temperature-Resistant - From -196°C to 100°C, Tough-Spots® withstand boiling water baths, moderate oven temperatures (150°C), autoclaving and extreme freezer temperatures (liquid and vapor phase nitrogen)
Specially Designed - Label sheets are specially top-coated to readily accept laser printing. Heat-resistant, stay-flat sheets will not jam in laser printers
Ideally Sized - Fits a variety of microcentrifuge tube tops, cryovial caps and lids, as well as many other laboratory container caps, lids and covers
Color Coding - 3/8 in. (9.5 mm) and 1/2 in. (13 mm) diameter Laser Tough-Spots® are available in White, Blue, Green, Orange, Red and Yellow
Caratteristiche e vantaggi
- Chemically inert, polyolefin labels resist most organic solvents and caustic agents.
- They adhere to most plastics†, glass, and metals without cracking, peeling, or degrading
- Temperature-Resistant from -196°C to 100°C
- Withstand boiling water baths, moderate oven temperatures (150°C), autoclaving and extreme freezer temperatures (liquid and vapor phase nitrogen)
- Label sheets are specially top-coated to readily accept laser printing. Heat-resistant, stay-flat sheets will not jam in laser printers
- Fits a variety of microcentrifuge tube tops, cryovial caps and lids, as well as many other laboratory container caps, lids and covers
Note legali
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
-
How many boxes will I receive when I choose quantity "1" of Z740259
1 answer-