Cambia visualizzazione
| Voi/SKU | Disponibilità | Prezzo |
|---|---|---|
12 ea | Per conoscere la disponibilità, visualizza il carrello | CHF 180.00 |
Informazioni su questo articolo
NACRES:
NB.22
UNSPSC Code:
41123100
CHF 180.00
Per conoscere la disponibilità, visualizza il carrello
Servizio Tecnico
Hai bisogno di aiuto? Il nostro team di scienziati qualificati è a tua disposizione.
Permettici di aiutartimaterial
blue
packaging
pack of 12 ea (dry with glycerol)
manufacturer/tradename
Spectrum Labs G235059
technique(s)
dialysis: suitable
pore size
100 kDa MWCO
capacity
5 mL
General description
- 95 - 98% Sample Recovery
- Highest Membrane Purity
- Superior Handling & Leak Prevention
- Volume Specific Dilution Control
- Complete & Ready-to-Use Packaging
Spectrum′s proprietary Biotech Grade CE is a low protein-binding synthetic membrane available in 4 precise MWCO′s (color coded for MWCO variants) with no heavy metal and sulfide contaminants. The cylindrical tubing geometry prevents sample dilution (associated with cassette-type devices) and provides open access for total volume retrieval. Only the Float-A-Lyzer® G2 assures a 95-98% sample recovery while maintaining 99% sample purity and <1% sample dilution.
The leak-proof screw-on cap with sealing o-ring provides easy access with included pipette for loading, in-process testing and sample retrieval, without the risk of needle punctures. The included floatation ring improves sample buoyancy and vertical orientation during dialysis. The sleek design allows multiple samples to be dialyzed in the same reservoir.
No additional kits, buoys or accessories are required. The Float-A-Lyzer G2 is packaged dry and ready-to-go with all components included. Simply rinse with water, load the sample with a pipette, and dialyze.
Spectrum Laboratories, Inc. introduces the Spectra/Por® Float-A-Lyzer® G2, the next generation of Ready-to-Use laboratory dialysis devices featuring a Ultra-pure Biotech Cellulose Ester (CE) Membrane
Legal Information
Float-A-Lyzer is a registered trademark of Spectrum Laboratories, Inc.
Spectra/Por is a registered trademark of Spectrum Laboratories, Inc.
1 of 1
Questo articolo | |||
|---|---|---|---|
| description blue, 5 mL, MWCO 100 kDa | description blue, 1 mL, MWCO 100 kDa | description yellow, 5 mL, MWCO 8-10 kDa | description red, 1 mL, MWCO 20 kDa |
| packaging pack of 12 ea (dry with glycerol) | packaging pack of 12 ea (dry with glycerol) | packaging pack of 12 ea (dry with glycerol) | packaging pack of 12 ea (dry with glycerol) |
| capacity 5 mL | capacity 1 mL | capacity 5 mL | capacity 1 mL |
| pore size 100 kDa MWCO | pore size 100 kDa MWCO | pore size 8-10 kDa MWCO | pore size 20 kDa MWCO |
| manufacturer/tradename Spectrum Labs G235059 | manufacturer/tradename Spectrum Labs G235035 | manufacturer/tradename Spectrum Labs G235055 | manufacturer/tradename Spectrum Labs G235033 |
| material blue | material blue | material yellow | material red |
Still not finding the right product?
Explore all of our products under Spectra-Por® Float-A-Lyzer® G2
Scegli una delle versioni più recenti:
Possiedi già questo prodotto?