Z722390
x-tracta Gel Extraction Tool
pkg of 100 ea
Sinonimo/i:
dna extraction, dna extraction tool, dna isolation, dna isolation tool, nucleic acid isolation, rna isolation, rna isolation tool
About This Item
Prodotti consigliati
Confezionamento
pkg of 100 ea
Descrizione generale
Benefits of the x-tracta Gel Extraction Tool:
- Fast
- East to Use
- One-Handed Procedure
- Precise Extraction
- High Yield Recovery
- Disposable
- Safe
For an instructional overview and further details regarding the x-tracta gel extraction tool, please visit www.sigmaaldrich.com/xtracta
Raccomandato
I clienti hanno visto anche
Protocolli
A quick, clean and easy to use alternative to using a scalpel or razor blade for the extraction of DNA and RNA bands from agarose gels following gel electrophoresis.
The GenElute Gel Extraction kit is designed for the rapid purification of 50 bp to 10 kb linear DNA fragments and plasmids from standard or low-melting agarose gels.