Documenti fondamentali
Z720046
Millipore
Millipore® Ziptips
C18, rack of 96
Sinonimo/i:
pipet tips, pipette tips
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.01
Prezzi e disponibilità al momento non sono disponibili
Materiali
colorless tip
Sterilità
non-sterile
Caratteristiche
barrier: no
Confezionamento
rack of 96
Produttore/marchio commerciale
Millipore ZTC18s960
Intervallo di volume
0.1-10 μL
Descrizione generale
Millipore ZipTips: Sample Preparation of Peptides or Proteins Prior to Electrospray/Nanoelectrospray MS
ZipTip Advantages:
  • Single-step desalting, concentration, and purification
  • Fractionate complex samples for more meaningful data
  • Ideal for peptides, proteins, nucleic acids, and more
  • No dead volume for maximum recovery
  • Eliminates time-consuming chromatography
Examples of analyses enhanced by ZipTip sample preparation:
  • Mass spectrometry
  • HPLC (high performance liquid chromatography)
  • Capillary electrophoresis
  • Atomic absorption spectroscopy
The ZipTip is a 10 μL (P-10) pipette tip with a bed of chromatography media fixed at its end such that there is no dead volume. It is intended for purifying and concentrating femtomoles to picomoles of protein, peptide or oligonucleotide samples prior to analysis, providing better data quality. The sample is aspirated and dispensed through ZipTip to bind, wash, and elute. Recovered samples are contaminant-free and eluted in 0.5-4 μL for direct transfer to a mass spectrometer target or vial.
ZipTip C18 is a 10 μL pipette tip with approximately a 0.6 μL bed of chromatography media fixed at its end such that there is no dead volume. ZipTip Micro-C18 is a 10 μL pipette tip with approximately a 0.2 μL bed of chromatography media fixed at its end such that there is no dead volume. Both are ideal for concentrating and purifying peptides, proteins or oligonucleotides in seconds prior to mass spectrometry, HPLC, capillary electrophoresis and other analytical techniques.
Note legali
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Arman Kulyyassov et al.
Biomolecules, 10(1) (2020-01-23)
Protein-protein interactions of core pluripotency transcription factors play an important role during cell reprogramming. Cell identity is controlled by a trio of transcription factors: Sox2, Oct4, and Nanog. Thus, methods that help to quantify protein-protein interactions may be useful for
Dae-Hee Lee et al.
Autophagy, 14(11), 1870-1885 (2018-07-07)
Macroautophagy is induced under various stresses to remove cytotoxic materials, including misfolded proteins and their aggregates. These protein cargoes are collected by specific autophagic receptors such as SQSTM1/p62 (sequestosome 1) and delivered to phagophores for lysosomal degradation. To date, little