Documenti fondamentali
Z711012
Media bottles, wide mouth, Lab 45, accessory
polypropylene pour ring
Sinonimo/i:
240756
Scegli un formato
12 EA
CHF 17.30
CHF 17.30
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
12 EA
CHF 17.30
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.17
CHF 17.30
Check Cart for Availability
Materiali
polypropylene pour ring
Produttore/marchio commerciale
WHEATON®
Caratteristiche e vantaggi
  • For high temperature applications, use the PBT (polybutylene terephthalate) screw caps and the ETFE (ethylene tetrafluoroethylene) pour lips which can withstand temperatures up to 200ºC up to 2 hours
  • Autoclavable
Note legali
WHEATON is a registered trademark of Wheaton Industries, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)