Z709832
Sartorius eLINE® multichannel electronic pipettor
volume 50-1200 μL, 12 channel with USA plug
Sinonimo/i:
Biohit pipettes, electronic pipettes, pipettor
About This Item
Codice UNSPSC:
41121500
Prodotti consigliati
Ingresso CA/CC
115 V AC
Caratteristiche
electronic
Produttore/marchio commerciale
Sartorius 730491
Disponibilità
not available in EU
Volume
50-1200 μL
Descrizione generale
With its easy-to-read display and simple-to-operate keyboard the hard work has been removed for programming Sartorius eLINE of the Biohitfamily. Boasting no fewer than 11 pipetting modes including:
The eLINE can store up to 6 user routines guaranteeing consistency of results irrelevant of the intricacy of the task undertaken. Tip ejection can be a chore at best and at worst a contributing factor to work-related upper limb disorders.
The eLINE negates this with its revolutionary electronic ejection system which discards the used tip at a touch of a button. Sartorius eLINE Electronic Pipettors should be charged using the original Sartorius eLINE Charging Stand. They also feature direct charging by using the Sartorius eLINE AC-adaptor.
- Forward pipetting with optional cycle counting
- Reverse and manual pipetting modes
- Multi-dispensing; Diluting
- Automated Dispensing
- Mixing facility
The eLINE can store up to 6 user routines guaranteeing consistency of results irrelevant of the intricacy of the task undertaken. Tip ejection can be a chore at best and at worst a contributing factor to work-related upper limb disorders.
The eLINE negates this with its revolutionary electronic ejection system which discards the used tip at a touch of a button. Sartorius eLINE Electronic Pipettors should be charged using the original Sartorius eLINE Charging Stand. They also feature direct charging by using the Sartorius eLINE AC-adaptor.
Note legali
eLINE is a registered trademark of Sartorius Biohit Liquid Handling Oy
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)