Documenti fondamentali
Z367788
Hi/Lo profile rocker accessories
two add-on platforms assembly
About This Item
Codice UNSPSC:
41100000
Produttore/marchio commerciale
IBI ACCRC00P2
Descrizione generale
Contains two accessory platforms for the Hi-Lo Rocker.
Kit includes all hardware necessary to add 2 accessory platforms to the Hi-Lo Rocker for a total of 3 platforms. Made of stainless steel for years of dependable service. Platforms can also be autoclaved to prevent contamination of incubators.
Made in the USA
Caratteristiche e vantaggi
  • Contains Two stainless steel platforms
  • All hardware included.
  • Transforms Hi-Lo Rocker into a Three platform unit.
Altre note
platform accessory for Z367745; Z367753
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)