Documenti fondamentali
Z363073
Sigma-Aldrich
Kodak® BioMax® MS film
Sinonimo/i:
autoradiography film, xray film
About This Item
Codice UNSPSC:
41105341
NACRES:
NB.22
Misura
8 in. (20 cm) × 10 in. (25 cm)
Descrizione generale
Double-emulsion film specifically engineered for biotechnology applications. Provides maximum sensitivity for detection of 32P and 125I in Northern and Southern blots, reducing quantity of isotope needed.
Each sheet of film is sealed in a light-tight envelope
Note legali
BioMax is a registered trademark of Carestream Health, Inc.
Kodak is a registered trademark of Eastman Kodak Company
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)