Documenti fondamentali
Z331716
BRAND® macro pipette controller
green
Sinonimo/i:
brand pipette, macro pipette controller
Scegli un formato
1 EA
CHF 72.90
CHF 72.90
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 72.90
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.01
CHF 72.90
Check Cart for Availability
Materiali
green
plastic (autoclavable)
silicone (autoclavable)
Caratteristiche
autoclavable
Produttore/marchio commerciale
BRAND 26201
Volume
0.1-200 mL
Categorie correlate
Descrizione generale
One single macro pipette controller covers the entire range of bulb and graduated pipettes from 0.1 to 200 mL. The conical silicone adapter offers a secure fit for the different diameters. The unit is fully autoclavable at 121°C (2 bar) according to DIN EN 285.
Supplied with a spare hydrophobic membrane filter which protects the system from liquid penetration.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)