Scegli un formato
| Voi/SKU | Disponibilità | Prezzo |
|---|---|---|
500 ea | Per conoscere la disponibilità, visualizza il carrello | CHF 457.00 |
Informazioni su questo articolo
Quality Segment
material
cap (urea), glass vial, metal foil liner
feature
closure type screw top vial, closure type white urea cap
packaging
pkg of 500 ea
vial size
28 mm × 61 mm , with cap
volume
20 mL
fitting
thread for 22-400
General description
Packaging
Caps are packed in sacks of 100 in separate trays.
1 of 1
Questo articolo | |||
|---|---|---|---|
| description volume 20 mL, vial size 28 mm × 61 mm , with cap, Cap: 22mm urea with metal foil liner. (caps packed separately), pkg of 500 ea | description volume 6 mL, Minivial, vial size 17 mm × 58 mm , with cap, Cap: 13mm urea with metal foil liner., pkg of 1000 ea | description volume 6 mL, Minivial, vial size 17 mm × 58 mm , with cap, Cap: 15mm urea with metal foil liner., pkg of 1000 ea | description volume 20 mL, vial size 28 mm × 61 mm , with cap, the cap is made of urea and the liner material is a polyethylene cone, pkg of 500 ea |
| material cap (urea), metal foil liner, glass vial | material metal foil liner, glass vial | material glass vial, metal foil liner | material glass vial |
| feature closure type screw top vial, closure type white urea cap | feature closure type white urea cap | feature closure type white urea cap | feature closure type polyethylene cap, poly-seal liner, closure type screw top vial |
| packaging pkg of 500 ea | packaging pkg of 1000 ea | packaging pkg of 1000 ea | packaging pkg of 500 ea |
| fitting thread for 22-400 | fitting thread for 13-425 | fitting thread for 15-425 | fitting thread for 22-400 |
| vial size 28 mm × 61 mm , with cap | vial size 17 mm × 58 mm , with cap | vial size 17 mm × 58 mm , with cap | vial size 28 mm × 61 mm , with cap |
| Quality Level 100 | Quality Level - | Quality Level 100 | Quality Level - |
Still not finding the right product?
Explore all of our products under Liquid scintillation vials, glass
Scegli una delle versioni più recenti:
Possiedi già questo prodotto?