Documenti fondamentali
TMODS2185-0032
Nalgene® economy wide-mouth bottle
amber high-density polyethylene, volume 1000 mL, case of 24 ea
Sinonimo/i:
amber HDPE bottle, nalgene HDPE bottle
Scegli un formato
24 EACH
CHF 309.00
CHF 309.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
24 EACH
CHF 309.00
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
CHF 309.00
Check Cart for Availability
Materiali
amber high-density polyethylene
Confezionamento
case of 24 ea
Produttore/marchio commerciale
Nalgene DS2185-0032
Capacità
1000 mL
Volume
1000 mL
Descrizione generale
Perform short-term general purpose laboratory and field applications with Thermo Scientific Nalgene Wide-Mouth HDPE Economy Bottles with Closure. These light-duty translucent bottles are a lightweight version of the original Nalgene bottle, bulk-packed for ultimate economy. Thinner-walled and more flexible than the Nalgene Lab Quality Bottles, they feature a wide mouth for easy filling and offer excellent chemical resistance to most acids, bases and alcohols.
Note legali
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)