Documenti fondamentali
TMO3665-1000
Nalgene® economy graduated cylinder
PMP, volume 1000 mL, case of 6 ea
Sinonimo/i:
PMP graduated cylinder, PMP graduated flask
Scegli un formato
6 EACH
CHF 224.00
CHF 224.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
6 EACH
CHF 224.00
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.43
CHF 224.00
Check Cart for Availability
Materiali
PMP
Confezionamento
case of 6 ea
Produttore/marchio commerciale
Nalgene 3665-1000
Capacità
1000 mL
Volume
1000 mL
Descrizione generale
Dispense liquids easily with simple to read, molded-in graduations that won′t rub off. Affordably priced Thermo Scientific Nalgene Polypropylene Economy Graduated Cylinders have a generous pour spout and exhibit excellent chemical and break resistance.
Note legali
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)