Documenti fondamentali
TMO3665-0100
Nalgene® economy graduated cylinder
PMP, volume 100 mL, case of 12 ea
Sinonimo/i:
PMP graduated cylinder, PMP graduated flask
Scegli un formato
12 EACH
CHF 194.00
CHF 194.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
12 EACH
CHF 194.00
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.43
CHF 194.00
Check Cart for Availability
Materiali
PMP
Confezionamento
case of 12 ea
Produttore/marchio commerciale
Nalgene 3665-0100
Capacità
100 mL
Volume
100 mL
Descrizione generale
Dispense liquids easily with simple to read, molded-in graduations that won′t rub off. Affordably priced Thermo Scientific Nalgene Polypropylene Economy Graduated Cylinders have a generous pour spout and exhibit excellent chemical and break resistance.
Note legali
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)