Documenti fondamentali
TMO362002-9025
Nalgene® diagnostic bottle without cap
natural high-density polyethylene, volume 8 mL, case of 2000 ea
Sinonimo/i:
PPCO bottles, small volume bottle
Scegli un formato
2000 EACH
CHF 1’030.00
CHF 1’030.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
2000 EACH
CHF 1’030.00
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
CHF 1’030.00
Check Cart for Availability
Materiali
natural high-density polyethylene
Confezionamento
case of 2000 ea
Produttore/marchio commerciale
Nalgene 362002-9025
Capacità
8 mL
Volume
8 mL
Descrizione generale
Safely store and ship light-sensitive diagnostic samples and valuable reagents in small volumes with Thermo Scientific Nalgene Opaque Amber HDPE Diagnostic Bottles, used in test kits and many small-volume applications. These opaque amber high-density polyethylene bottles are for use with light sensitive reagents and are packed without closures so they can be matched with various colored closures (Cat. No. 362150 series).
Note legali
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)