Documenti fondamentali
TMO331827-BR
Nunc® barcoded latch rack
for 1.0-2.0 mL cryogenic tubes, plastic, 42 positions, case of 10 ea
Sinonimo/i:
tube rack, tube storage
Scegli un formato
10 EACH
CHF 278.00
CHF 278.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
10 EACH
CHF 278.00
About This Item
Codice UNSPSC:
41122804
NACRES:
NB.12
CHF 278.00
Check Cart for Availability
Materiali
plastic
Confezionamento
case of 10 ea
Produttore/marchio commerciale
Nunc 331827-BR
Descrizione generale
Store tubes for automated picking/placing, liquid handling and other high throughput applications using the Thermo Scientific Nunc Universal Latch Rack for Cryogenic Tubes.
Use versatile Thermo Scientific Nunc 1.8 mL Externally-threaded Universal Tubes either inside a large storage system, or in smaller scale when used for traditional cryostorage in dewars. Available racked or in bulk, with 2D barcodes or non-coded.
Note legali
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)