Documenti fondamentali
TMO3103-0015
Nalgene® centrifuge tubes
conical bottom clear polypropylene copolymer, volume 15 mL, without cap, non-sterile, case of 100 ea
Sinonimo/i:
Graduated Conical Bottom Centrifuge Tubes
Scegli un formato
100 EACH
CHF 757.00
CHF 757.00
Spedizione prevista il26 maggio 2025
Scegli un formato
Cambia visualizzazione
100 EACH
CHF 757.00
About This Item
Codice UNSPSC:
41121703
NACRES:
NB.11
CHF 757.00
Spedizione prevista il26 maggio 2025
Materiali
conical bottom clear polypropylene copolymer
Sterilità
non-sterile
Caratteristiche
without cap
Confezionamento
case of 100 ea
Produttore/marchio commerciale
Nalgene 3103-0015
Volume
15 mL
Descrizione generale
Simplify measurements using Thermo Scientific Nalgene PPCO Conical-Bottom Centrifuge Tubes, which feature molded-in graduations. Translucent polypropylene copolymer (PPCO) has excellent chemical resistance.
<UL><LI>Translucent Polypropylene Copolymer (PPCO) is compatible with a wide range of lab reagents</LI><LI>Designed for low-speed centrifugation up to 6000 × g in refrigerated or non-refrigerated centrifuges (Tubes must be filled at least 80% for proper performance)</LI><LI>Conical bottoms concentrate pellet in a small area for easy isolation and retrieval</LI><LI>Molded-in graduations last the life of the tube</LI><LI>Last longer than polycarbonate tubes under conditions of repeated autoclaving</LI><LI>Tube are both autoclavable and graduated</LI></UL>
Note legali
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)