Documenti fondamentali
TMO167554
Nunc® Edge 2.0 96-well Plate
Nunclon® Delta surface, polystyrene, sterile, without lid, clear, case of 160 ea
Sinonimo/i:
cell culture plate, cell growth plate
Scegli un formato
160 EACH
CHF 405.00
CHF 405.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
160 EACH
CHF 405.00
About This Item
Codice UNSPSC:
41122102
NACRES:
NB.15
CHF 405.00
Check Cart for Availability
Materiali
clear
polystyrene
Sterilità
sterile
Confezionamento
case of 160 ea
Produttore/marchio commerciale
Nunc 167554
Descrizione generale
Minimize plate evaporation and the risks associated with edge effect by using the uniquely engineered Thermo Scientific Nunc Edge 2.0 Plate. The surrounding bulit-in reservoirs, when filled with sterile water or media, act as evaporation barriers, yielding consistency across the entire 96-well plate. Enabling the use of all 96 wells in a plate allows researchers to plan experiments more efficiently with enhanced productivity and less waste for cell-based assays that require extended incubation.
Note legali
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)