Merck
R0151
NovaRack
pink
Sinonimo/i:
Diversified Biotech NORC-4000, numbered tube rack, rube rack, tube holder, tube storage
Autenticati per visualizzare i prezzi organizzativi e contrattuali.
Scegli un formato
Cambia visualizzazione
Informazioni su questo articolo
NACRES:
NB.35
UNSPSC Code:
41122804
Manufacturer/tradename:
Diversified Biotech NORC-4000
Feature:
Holds 1 x 50 mL tubes, Holds 14 x 13 mm tubes, Holds 24 x 0.5-2 mL tubes, Holds 7 x 15 mm tubes
Packaging:
pack of 1 ea
Servizio Tecnico
Hai bisogno di aiuto? Il nostro team di scienziati qualificati è a tua disposizione.
Permettici di aiutarti
feature
Holds 1 x 50 mL tubes, Holds 14 x 13 mm tubes, Holds 24 x 0.5-2 mL tubes, Holds 7 x 15 mm tubes
packaging
pack of 1 ea
manufacturer/tradename
Diversified Biotech NORC-4000
color
pink
General description
NovaRack provides universal housing for test tubes and vials of different sizes on the same surface. The numeric labeling for each tube cavity allows one to easily organize and track up to 46 tubes. It accommodates 24 microcentrifuge tubes (0.5-2 mL), 14 test tubes (12 x 75 mm culture tubes, cryovials and a variety of blood collection tubes), 7 centrifuge tubes (15 mL) and one 50 mL tube. NovaRack is constructed of polypropylene, is autoclavable and is available in five colors. With the addition of the Carousel Extension rod, the space saving ability is doubled and the top rack is allowed to spin.
Legal Information
Novarack is a trademark of Diversified Biotech
Still not finding the right product?
Explore all of our products under NovaRack
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)