Documenti fondamentali
OH30253005
Ohaus® Scout® Portable Touchscreen Balance
Model STX123, weighing capacity 120 g, resolution: 0.001 g, AC/DC input 110 V, US 3-pin plug (North America)
About This Item
Codice UNSPSC:
41111502
Materiali
metal weighing plate
Caratteristiche
CSA approved
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Ohaus® 30253005
Capacità di pesata
120 g, resolution: 0.001 g
Diam. piattino
9.3 cm
Descrizione generale
Ideal for laboratory and industrial applications, the OHAUS Scout features a user-friendly color touchscreen and a compact stackable design. Designed to deliver superior performance with rapid stability testing and high resolution weighing results, these portable scales set a new standard in weighing applications. Their features also include superior overload protection and advanced connectivity options.
Applicazioni
Weighing, piece counting,% weighing, check weighing, animal weighing / dynamic weighing, totalizing, density determination, display hold, mole weighing
Caratteristiche e vantaggi
Stability indicator, stackable design with storage cover included (except 1 mg models), mechanical and software overload / underload protection, low battery level indicator, automatic shutdown, auto-tare, slope calibration mass included with models up to 620g.
Note legali
Ohaus is a registered trademark of Ohaus Corp.
Scout is a registered trademark of Ohaus Corp.