MTCP5200300U
MTC™ Bio ProPette™ LE Adjustable Pipettor
capacity 30-300 μL
Sinonimo/i:
pipette
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.15
Prodotti consigliati
Caratteristiche
adjustable
manual
no. of channels 1
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
MTC™ Bio P5200-300U
Capacità
30-300 μL
Descrizione generale
ProPette LE™ Single-Channel Pipettes provide maximum precision at virtually every conceivable volume. They feature a digital micrometer that is adjustable for precise volume control as well as a mechanical volume lock positioned at the top of the pipette that can be engaged or disengaged with one hand. These pipettors are lightweight and finely balanced for maximum ergonomics.
ProPette™ LE, 30 to 300µL
Caratteristiche e vantaggi
- Adjustable with Roto-Lock
- Light weight with reduced forces
- Full volume range (0.1µL-10mL)
- Superior accuracy and reproducibility
- Outstanding user comfort
Note legali
MTC is a trademark of MTC Bio
ProPette is a trademark of MTC Bio
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)