MTCP4300RK
MTC™ Bio Non-Filtered Pipette Tips
capacity 300 μL, sterile, rack of 10 × 96 ea
Sinonimo/i:
pipetting tip, plastic tube, sample storage tube
About This Item
Prodotti consigliati
Materiali
clear
polypropylene
Sterilità
sterile
Caratteristiche
DNase free
RNase free
Confezionamento
rack of 10 × 96 ea
Produttore/marchio commerciale
MTC™ Bio P4300-RK
Capacità
300 μL
Categorie correlate
Descrizione generale
Pipette Tips, Non-filtered, 300µL capacity, sterile, for 200µL and 300µL pipettes, 10 racks of 96 tips
Caratteristiche e vantaggi
- For 300µL pipettes
- Also fits most 200µL pipettes
- Filtered or non-filtered
- Pre-sterilized
- Autoclavable box-rack
Note legali
MTC is a trademark of MTC Bio
ProPette is a trademark of MTC Bio