Documenti
M1435
Sigma-Aldrich
Microarray Spotting Solution for Silane Coated Slides
About This Item
Codice UNSPSC:
12352200
Caratteristiche e vantaggi
Improves spot morphology and DNA binding to the slide resulting in increased hybridization signal.
Altre note
Optimized spotting solution for use with SigmaScreen™ APS Coated Slides for Microarrays (Product Code S9936).