Documenti fondamentali
L6414
Sigma-Aldrich
Laser Tough-Spots®
diam. 1/2 in., red
Sinonimo/i:
solvent resistant labels, temperature resistant labels
Scegli un formato
1 PKG
CHF 77.50
CHF 77.50
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 PKG
CHF 77.50
About This Item
Codice UNSPSC:
55121611
NACRES:
NB.35
CHF 77.50
Check Cart for Availability
Materiali
polyolefin
red
Confezionamento
pack of 3840 ea (192 labels per sheet)
Produttore/marchio commerciale
Diversified Biotech SPOT-2400
Diametro
1/2 in.
Categorie correlate
Descrizione generale
Tough-Spots are chemically inert, polyvinyl labels that adhere to all plastics. Tough-Spots withstand boiling water baths, conventional and cryogenic freezer temperatures, most organic solvents, caustic agents and other challenges without peeling, becoming brittle or degrading. Labels can be printed directly from the laser printer. 20 sheets per pack.
Note legali
Tough-Spots is a registered trademark of Diversified Biotech
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)