Vai a
HS15989E
Premium slide box
25 place, cork lining, white plastic
Sinonimo/i:
Schiebekasten, slide box microscope, slide storage, slide storage box
Autenticati per visualizzare i prezzi organizzativi e contrattuali.
Scegli un formato
Cambia visualizzazione
Voi/SKUDisponibilitàPrezzo
Informazioni su questo articolo
NACRES:
NB.17
UNSPSC Code:
41123302
Manufacturer/tradename:
Heathrow Scientific HS15989E
Feature:
Holds 25 x slides 76x26 mm
Material:
nickel closure, white plastic
Prezzi e disponibilità al momento non sono disponibili
Servizio Tecnico
Hai bisogno di aiuto? Il nostro team di scienziati qualificati è a tua disposizione.
Permettici di aiutarti
material
nickel closure, white plastic
feature
Holds 25 x slides 76x26 mm
manufacturer/tradename
Heathrow Scientific HS15989E
L × W × H
141 × 88 × 35 mm
Features and Benefits
Slide boxes are perfect for storage or transportation of standard size slides (100 (50 back to back) x 75 mm and 1 x 3").
  • Molded, rigid boxes securely hold slides in grooved slots
  • Durable ABS plastic boxes are available with cork lining only
  • Both the lining and the slots are numbered to correspond to the slide inventory sheet
  • Feature thumb-latch lock mechanism
Confronta articoli simili
Visualizza il confronto per intero
Mostra le differenze
1 of 1
Questo articolo
HS15989RHS15989BHS15989M
Premium slide box 25 place, cork lining, white plastic
HS15989E
Premium slide box
Premium slide box 25 place, foam lining, white plastic
HS15989R
Premium slide box
Premium slide box 25 place, cork lining, green plastic
HS15989B
Premium slide box
Premium slide box 25 place, foam lining, blue plastic
HS15989M
Premium slide box
description
25 place, cork lining, white plastic
description
25 place, foam lining, white plastic
description
25 place, cork lining, green plastic
description
25 place, foam lining, blue plastic
feature
Holds 25 x slides 76x26 mm
feature
Holds 25 x slides 76x26 mm
feature
Holds 25 x slides 76x26 mm
feature
Holds 25 x slides 76x26 mm
manufacturer/tradename
Heathrow Scientific HS15989E
manufacturer/tradename
Heathrow Scientific HS15989R
manufacturer/tradename
Heathrow Scientific HS15989B
manufacturer/tradename
Heathrow Scientific HS15989M
material
nickel closure, white plastic
material
nickel closure, white plastic
material
nickel closure, green plastic
material
blue plastic , nickel closure
L × W × H
141 × 88 × 35 mm
L × W × H
141 × 88 × 35 mm
L × W × H
141 × 88 × 35 mm
L × W × H
141 × 88 × 35 mm
Still not finding the right product?
Explore all of our products under Premium slide box
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)