Documenti fondamentali
HS1421A
Weigh Boat
Small, square blue polystyrene, pk of 500 ea
Sinonimo/i:
weigh boat, weigh dish, weighing dish, weighing pan
About This Item
Codice UNSPSC:
41121812
NACRES:
NB.35
Prezzi e disponibilità al momento non sono disponibili
Materiali
square blue polystyrene
Misura
small
Confezionamento
pk of 500 ea
Produttore/marchio commerciale
Heathrow Scientific HS1421A
Descrizione generale
Flexible polystyrene boats feature smooth surfaces to provide accurate pour-outs with minimal sample loss.
Caratteristiche e vantaggi
  • biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases.
  • temperature range -10 to 70C
  • disposable
  • shallow with a wide, flat bottom to resist tipping
  • rounded corners simplify transfer
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)