Documenti fondamentali
HS120204
Microtube Storage Box
orange polypropylene, pk of 5, Holds 100 x 1.5-2.0 mL tubes
About This Item
Codice UNSPSC:
41123300
NACRES:
NB.22
Prezzi e disponibilità al momento non sono disponibili
Materiali
orange polypropylene
Caratteristiche
Holds 100 x 1.5-2.0 mL tubes
Confezionamento
pk of 5
Produttore/marchio commerciale
Heathrow Scientific HS120204
Diam.pozzetto
12.3 mm
Caratteristiche e vantaggi
  • for larger storage requirements of 1.5-2.0 mL microtubes or cryogenic vials
  • easy-open hinged lid with snap clasp
  • autoclavable
  • designed for quick visual identification
  • uniform wells with location indentification
  • tube capacity is indicated on the outside of the box
  • translucent vibrant colors with frosted labeling area
  • debossed with a distinctive wave design
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)