GN782101
Greiner microplate
1536 well, HiBase, flat bottom, clear
Sinonimo/i:
1536 well microplate, 1536 well plate, cell culture plate
About This Item
Codice UNSPSC:
41121800
Materiali
flat bottom
Confezionamento
pack of 60 ea
Produttore/marchio commerciale
Greiner 782101
Disponibilità
available only in China
Misura
1536 wells
Compatibilità
suitable for (cell culture)
Descrizione generale
- Greiner microplates are ideal for storing active agents, patient samples, DNA/RNA or stock cultures.
- Polypropylene material can withstand a high temperature (-196 to +121°C) and are chemically resistant.
- Nonpyrogenic plates are free of detectable DNase, RNase, human DNA.
- Features alphanumeric well coding for identification.
- Sealable with adhesive films and heat sealers for long-term and secure sealing.
Note legali
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)