Documenti fondamentali
GN781287
Greiner microplate
384 well, conical bottom, white
Sinonimo/i:
384 well microplate, 384 well plate, cell culture plate
About This Item
Codice UNSPSC:
41121800
Materiali
white
Confezionamento
pack of 100 ea
Produttore/marchio commerciale
Greiner 781287
Disponibilità
available only in China
Misura
384 wells
Compatibilità
suitable for (cell culture)
Descrizione generale
  • Greiner microplates are ideal for storing active agents, patient samples, DNA/RNA or stock cultures.
  • Polypropylene material can withstand a high temperature (-196 to +121°C) and are chemically resistant.
  • Nonpyrogenic plates are free of detectable DNase, RNase, human DNA.
  • Features alphanumeric well coding for identification.
  • Sealable with adhesive films and heat sealers for long-term and secure sealing.
Note legali
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)