Documenti fondamentali
GEPR648
Slot Blot Manifold
PR648, pack of 1 ea
About This Item
Codice UNSPSC:
41100000
NACRES:
NB.51
Materiali
acrylic plate
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Cytiva 80609558
Descrizione generale
PR 648 slot blot filtration manifolds are designed for ease of use and maximum filtration with minimum chance of cross-contamination. They make screening of many samples quick and easy. PR 648 Slot Blot Filtration Manifolds are designed for reproducibility. Large screws and rim design produce well-defined slots and prevent sample leakage between wells. Up to 1 ml can be filtered onto the 6.0 × 0.8 mm slot area.
Caratteristiche e vantaggi
  • Filter DNA, RNA, or protein into clearly defined slots on nitrocellulose or nylon membranes.
  • Clamp membranes securely with raised well edges to eliminate cross-contamination without O-rings or gaskets.
  • Transfer samples from microplates to wells with standard multichannel pipettors.
  • Track sample locations with numbered slots and notched membrane shape.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)