GE11-0007-68
Solvent Filter
(PTFE), Cytiva
About This Item
Materiali
(PTFE)
Descrizione
The wetted parts are resistant to organic solvents and salt buffers commonly used in chromatography of biomolecules, except 100% Ethyl acetate, 100% Hexane and 100% Tetrahydrofuran
Caratteristiche
Wetted Material: Polytetrafluoroethene (PTFE)
Produttore/marchio commerciale
11-0007-68
Cytiva
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)