GE11-0002-93
Ettan™ spot picker non-backed gel kit
Sinonimo/i:
11-0002-93, protein handling
About This Item
Codice UNSPSC:
41100000
Produttore/marchio commerciale
Cytiva 11-0002-93
Cytiva 11000293 (Article Number)
Descrizione generale
- Allows quick and accurate protein spot picking from nonbacked 2-D gels, increasing the versatility of Ettan Spot Picker.
- Compatible with common staining methods such as silver, Coomassie Blue, fluorescence, and labeling with Ettan DIGE Fluor dyes.
- High-precision picking with reference markers - using the gel frame, a nonbacked gel slab is attached to a temporary cellophane backing, supporting the reference markers.
- Improves nonbacked gel handling during scanning, and the glass lid protects the moist gel.
Kit Contains:
- 1 x Picking platform
- 1 x Gel frame (outer frame and inner frame)
- 1 x Gel loading platform
- 50 x Porous cellophane
- 2 x Glass lid for gel carrier
Note legali
Ettan is a trademark of Cytiva
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)