Documenti fondamentali
GBL246824
Grace Bio-Labs ProPlate® microarray system tray set
well size 24, Square, With Stainless Steel Spring Clips
Sinonimo/i:
incubation chambers, microarray hybridization chambers, slide chambers
Scegli un formato
1 EA
CHF 250.00
CHF 250.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 250.00
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.17
CHF 250.00
Check Cart for Availability
Materiali
black silicone (Acrylic tray)
Caratteristiche
Includes Tray and Cover, 10 Seal Strips, and 1 Applicator
Square, With Stainless Steel Spring Clips
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Grace Bio-Labs 246824
Dimensione pozzetto
24
Descrizione generale
ProPlate re-usable, multi-well chambers form removable, leak-proof wells on virtually any slide surface without the use of adhesive.
Designs feature ANSI-SLAS compliant well spacing to facilitate multichannel pipetting.
Multiple slides insert easily into frames having a standard microtiter plate footprint for automated processing.
Note legali
ProPlate is a registered trademark of Grace Bio-Labs, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)