Documenti
EP5427739009
Eppendorf® Fixed-angle rotor
F-35-6-30, incl. rotor lid
About This Item
Codice UNSPSC:
41103909
NACRES:
NB.82
Caratteristiche
to hold 12 x Vacutainer® tubes
to hold 6 x 15/50 mL tubes
F-35-6-30, incl. rotor lid
Confezionamento
pack of 1 ea
pkg of 1 ea
Produttore/marchio commerciale
Eppendorf® 5427739009
Parametri
7,830 max. RCF (7,745 xg)
tecniche
centrifugation: suitable
Compatibilità
for use with centrifuge models 5430/5430R
Note legali
Eppendorf is a registered trademark of Eppendorf AG