Documenti
EP5424706002
Eppendorf® Rotor F-45-18-11-Kit
incl.lid
About This Item
Codice UNSPSC:
41103909
NACRES:
NB.82
Caratteristiche
to hold 18 x 1.5/2.0 mL tubes
to hold 18 x spin columns
incl.lid
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Eppendorf® 5424706002
Parametri
15,000 max. RCF (18,111 xg)
tecniche
centrifugation: suitable
Compatibilità
for use with centrifuge models 5424/5424R
Note legali
Eppendorf is a registered trademark of Eppendorf AG