EP4861611006
Eppendorf® Pipette lower part
12-channel, size 15-300 μL, orange
About This Item
Caratteristiche
12-channel
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Eppendorf® 4861611006
Misura
15-300 μL
Colore
orange
Note legali
Eppendorf is a registered trademark of Eppendorf AG