Documenti fondamentali
EP3123000900
Eppendorf Research® Plus G
3-pack, Option 1, 0.5-10 uL/10-100 uL/100-1000 uL, includes epT.I.P.S.® box
Sinonimo/i:
Adjustable volume pipette
Scegli un formato
1 EA
CHF 846.00
CHF 846.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 846.00
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.01
CHF 846.00
Check Cart for Availability
Caratteristiche
single-channel
3-pack, Option 1, 0.5-10 uL/10-100 uL/100-1000 uL, includes epT.I.P.S.® box
Confezionamento
pack of 1 kit
Produttore/marchio commerciale
Eppendorf® 3123000900
Volume
× 10 μL
× 100 μL
× 1000 μL
Descrizione generale
The Eppendorf Research plus mechanical pipette combines over 50 years of innovation in liquid handling and is one of the most commonly used pipettes in the world. It provides you with one of the safest and most ergonomic pipettes available today. The Research plus microliter pipette protects the health of our customers during their daily work in the laboratory. The Research plus is the pipette family with the lowest weight and lowest operation forces in the Eppendorf product families. Lower operation forces can only be reached with our Eppendorf electronic pipettes. Enjoy excellent flexibility and choose among single-channel pipettes in fixed or variable volume as well as multi-channel pipette options with 8, 12, 16 and 24 channels.
Product features
It is a good feeling to know you are doing the right thing – to know you are working with one of the most advanced pipettes in the world. A spring loaded tip cone, a secondary calibration option, an improved volume display – and all that in an ultra light, fully autoclavable pipette: That is the Eppendorf Research plus pipette.
Accoladed product
The Research plus mechanical pipette was accoladed with the Bronze “Seal of Quality”. These seals are given to the top 0.1 % of products used in laboratories that have continually received the highest user review ratings. The award is praised as providing “invaluable” insights for scientists and intends to help them choose the ideal laboratory product for their working environment.
Note legali
Eppendorf is a registered trademark of Eppendorf AG
Eppendorf Research is a registered trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)