Documenti fondamentali
EP0030140524
Storage Box for Eppendorf® Tubes
size 8 × 8, pack of 3 ×, Holds 64 x 1-2 mL tubes
Scegli un formato
3 EA
CHF 34.80
CHF 34.80
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
3 EA
CHF 34.80
About This Item
Codice UNSPSC:
41123300
NACRES:
NB.12
CHF 34.80
Check Cart for Availability
Materiali
polypropylene
Caratteristiche
Holds 64 x 1-2 mL tubes
Produttore/marchio commerciale
Eppendorf® 0030140524 (Global)
Confezionamento
pack of 3 ×
Altezza
53 mm
Misura
8 × 8
Descrizione generale
Storage Box 8 x 8, for 64 tubes, 3 pcs., height 52.8 mm, 2 inch, polypropylene, for freezing to -86 °C, autoclavable, with lid and alphanumeric code
The comprehensive modular concept allows selection of the optimal freezer box combinations for individual storage demands. The outer footprint dimensions (133 mm) of these freezer boxes are compatible with common freezer rack systems and the different inner grid variants provide for an optimal fit to all typical tube formats: cryogenic tubes, microcentrifuge tubes, 15, 25 and 50 mL conical tubes and other laboratory vessels. This space-saving approach together with the application benefits resulting from the listed product features help to optimize your longterm storage and archiving of samples.
Note legali
Eppendorf is a registered trademark of Eppendorf AG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)